Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD38 Ab-3, Mouse Monoclonal Antibody

Catalog No. MS983R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS983R1 1 mL Prediluted
MS983S1 500 μL Concentrated
MS983S 1 mL Concentrated
MS983R7 7 mL Prediluted
MS983S0 100 μL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS983R1 Supplier Epredia™ Supplier No. MS983R1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD38 Ab-3, Mouse Monoclonal Antibody.

CD38 is a single chain type II integral transmembrane protein, which is highly expressed on thymocytes. It is also present on activated T cells and terminally differentiated B cells (plasma cells). Other reactive cells include NK cells, monocytes, macrophages, and dendritic cells. CD38 may be detected on cells from multiple myeloma, ALL (B and T) and some AML. It is, however, not found on most mature resting peripheral lymphocytes. CD38 functions as a multicatalytic ectoenzyme serving as ADP-ribosyl cyclase, cyclic ADP-ribose hydrolase and possibly NAD+ glycohydrolase or as a cell surface receptor. Antibodies to CD38 are useful in subtyping of lymphomas and leukemias, detection of plasma cells (i.e. identification of myelomas), and as a marker for activated B and T cells.

Host Species: Mouse

Clone: 38C03; same as SPC32

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Extracellular

Immunogen: Recombinant protein encoding the extracellular domain of human CD38

Molecular Weight: 45 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD38 Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 38C03; same as SPC32
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the extracellular domain of human CD38
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.