Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD42b (GPIb) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1174R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1174R1 1 mL Ready-to-Use Prediluted
MS1174P1 500 μL Concentrated 200μg/mL
MS1174P 1 mL Concentrated 200μg/mL
MS1174P0 100 μL Concentrated 200μg/mL
MS1174P1ABX 100 μL Concentrated 1mg/mL
MS1174PABX 200 μL Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1174R1 Supplier Epredia™ Supplier No. MS1174R1

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.

The CD42b glycoprotein, also known as GPIb, is a cofactor of ristocetin-induced aggregation and is involved in the binding of platelets to blood vessel walls. The CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. The absence of CD42b antigen on platelets may indicate Bernard-Soulier disease.

Host Species: Mouse

Clone: 42C01

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant CD42b.

Molecular Weight: 135-145 kDa (α-chain) and 22-25 kDa (β-chain)

Positive Control: Tonsil

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD42b Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 42C01
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Recombinant CD42b
Quantity 1 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Ready-to-Use
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.