Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43/T-Cell Marker Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS146P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS146P1 500 μL Concentrated 200μg/mL
MS146P0 100 μL Concentrated 200μg/mL
MS146P1ABX 100 μL Concentrated 1mg/mL
MS146PABX 200 μL Concentrated 1mg/mL
MS146P 1 mL Concentrated 200μg/mL
MS146R1 1 mL Prediluted Prediluted
MS146RQ 12 mL Ready-to-Use Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS146P1 Supplier Epredia Supplier No. MS146P1

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry applications with Epredia™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: DF-T1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested

Epitope: Not determined

Immunogen: Myeloblastic KG1 cells

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Specifications

Antigen CD43/T-Cell Marker Ab-1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DF-T1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Myeloblastic KG1 cells
Quantity 500 μL
Regulatory Status IVD
Research Discipline T-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.