Learn More
Description
Host Species: Mouse
Clone: DF-T1
Isotype: IgG1
Species Reactivity: Human. Does not react with rat. Others not tested
Epitope: Not determined
Immunogen: Myeloblastic KG1 cells
Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Specifications
Specifications
| Antigen | CD43/T-Cell Marker Ab-1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | DF-T1 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Myeloblastic KG1 cells |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.