Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45/T200/LCA Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS355P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS355P0 100 μL Concentrated 200μg/mL
MS355P1 500 μL Concentrated 200μg/mL
MS355P 1 mL Concentrated 200μg/mL
MS355R7 7 mL Prediluted Prediluted
MS355RQ 12 mL Ready-to-Use Prediluted
MS355P1ABX 100 μL Concentrated 1mg/mL
MS355PABX 200 μL Concentrated 1mg/mL
MS355R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS355P0 Supplier Epredia Supplier No. MS355P0

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments.

CD45 leucocyte common antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals. Antibody to CD45 is useful in differential identification of lymphoid tumors from non-hematopoietic undifferentiated neoplasms.

Host Species: Mouse

Clone: PD7/26/16 + 2B11

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat and dog. Others not known

Epitope: Not determined

Immunogen: PD7/26/16: human peripheral blood lymphocytes maintained in T-cell growth factor. 2B11: isolated neoplastic cells from T-cell lymphoma

Molecular Weight: 220, 205, 190, 180 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Specifications

Antigen CD45/T200/LCA Ab-2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PD7/26/16 + 2B11
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen PD7/26/16: human peripheral blood lymphocytes maintained in T cell growth factor. 2B11: isolated neoplastic cells from T cell lymphoma.
Quantity 100 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.