Learn More
Description
Host Species: Mouse
Clone: UCHL-1
Isotype: IgG2a/κ
Species Reactivity: Human, Rhesus Monkey and Chimpanzee. Does not react with rat. Others not known
Epitope: Not determined
Immunogen: Interleukin-2-dependent human T lymphocytes
Molecular Weight: 180 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Specifications
Specifications
| Antigen | CD45RO (T-Cell Marker) Ab-1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | UCHL-1 |
| Concentration | 100ug/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Interleukin-2-dependent human T lymphocytes |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.