Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RO (T-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS112PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
200 μL
5 Slides
500 μL
Form:
Concentrated
Liquid
Prediluted
Concentration:
100ug/mL
1mg/mL
N/A
Prediluted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS112PABX 200 μL Concentrated 1mg/mL
MS112P1 500 μL Concentrated 100ug/mL
MS112PCS 5 Slides Liquid N/A
MS112P0 100 μL Concentrated 100ug/mL
MS112P1ABX 100 μL Concentrated 1mg/mL
MS112R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS112PABX Supplier Epredia Supplier No. MS112PABX
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Epredia™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody.

CD45RO (an isoform of leukocyte common antigen) is expressed on most thymocytes, monocytes, macrophages and granulocytes. It is not expressed on normal B lymphocytes. Intrathymic T cell maturation progresses from CD45RA-/CD45RO+ to CD45RA+/CD45RO-; while peripheral T cell maturation progresses from CD45RA+/CD45RO- to CD45RA-/CD45RO+.

Host Species: Mouse

Clone: UCHL-1

Isotype: IgG2a/κ

Species Reactivity: Human, Rhesus Monkey and Chimpanzee. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Interleukin-2-dependent human T lymphocytes

Molecular Weight: 180 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Specifications

Antigen CD45RO (T-Cell Marker) Ab-1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UCHL-1
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Interleukin-2-dependent human T lymphocytes
Quantity 200 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Non-human Primate
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.