Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1302P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1302P 1 mL 200μg/mL
MS1302PABX 200 μL 1mg/mL
MS1302P1 500 μL 200μg/mL
MS1302P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1302P Supplier Epredia Supplier No. MS1302P

Ensure accurate, reproducible results in immunofluorescence, immunoprecipitation and flow cytometry experiments with Epredia™ CD47 Ab-2, Mouse Monoclonal Antibody.

CD47 antigen, also known as integrin associated-protein (IAP), is expressed on all hematopoietic cells, including leukocytes, platelets and erythrocytes. It is also expressed on epithelial cells, endothelial cells, fibroblasts and many tumor cell lines. CD47 may play a role as a signal transducer in the regulation of cation fluxes across cell membranes and in the chemotactic and adhesive interactions of leukocytes with endothelial cells.

Host Species: Mouse

Clone: B6H12.2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Ig domain

Immunogen: Intact CD47 purified from placenta

Molecular Weight: 47-52 kDa (reduced) and 45-50.5, 110 kDa (nonreduced)

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence (Methanol fixed only)
  • Immunoprecipitation (Native only)
  • Flow Cytometry
  • Blocks binding of SIRPα
  • Inhibits in assays in which CD47-integrin association is required

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD47 Ab-2
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone B6H12.2
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Intact CD47 purified from placenta
Quantity 1 mL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.