Learn More
Description
Host Species: Mouse
Clone: B6H12.2
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Ig domain
Immunogen: Intact CD47 purified from placenta
Molecular Weight: 47-52 kDa (reduced) and 45-50.5, 110 kDa (nonreduced)
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence (Methanol fixed only)
- Immunoprecipitation (Native only)
- Flow Cytometry
- Blocks binding of SIRPα
- Inhibits in assays in which CD47-integrin association is required
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD47 Ab-2 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | B6H12.2 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Intact CD47 purified from placenta |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.