Learn More
Description
CD5, a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphoma, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.
Host Species: Mouse
Clone: 4C7
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: External domain
Immunogen: Recombinant protein corresponding to the external domain of the CD5 molecule
Molecular Weight: 67kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Specifications
Specifications
| Antigen | CD5 Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 4C7 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to the external domain of the CD5 molecule |
| Quantity | 7 mL |
| Regulatory Status | IVD |
| Research Discipline | Hematopoietic |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.