Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD5 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS393S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS393S1 500 μL Concentrated
MS393S 1 mL Concentrated
MS393RQ 12 mL Ready-to-Use
MS393S0 100 μL Concentrated
MS393R1 1 mL Prediluted
MS393R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS393S1 Supplier Epredia™ Supplier No. MS393S1

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD5 Ab-1, Mouse Monoclonal Antibody.

CD5, a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphoma, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.

Host Species: Mouse

Clone: 4C7

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: External domain

Immunogen: Recombinant protein corresponding to the external domain of the CD5 molecule

Molecular Weight: 67kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Specifications

Antigen CD5 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C7
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to the external domain of the CD5 molecule
Quantity 500 μL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.