Learn More
Description
Host Species: Rabbit
Clone: SP19
Species Reactivity: Human. Others not tested.
Epitope: Intracellular region
Immunogen: Synthetic peptide from the intracellular region of CD5
Positive Control: Raji cells, Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Specifications
| Antigen | CD5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SP19 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from the intracellular region of CD5 |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Hematopoietic |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.