Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Rabbit Monoclonal Antibody, Epredia™

Catalog No. RM9119R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9119R7 7 mL Prediluted
RM9119S1 500 μL Concentrated
RM9119S 1 mL Concentrated
RM9119R1 1 mL Prediluted
RM9119RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM9119R7 Supplier Epredia Supplier No. RM9119R7

Obtain accurate and reproducible results in immunohistochemistry and western blotting procedures with Epredia™ CD5, Rabbit Monoclonal Antibody.

CD5, a transmembrane protein, is found on 95%of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.

Host Species: Rabbit

Clone: SP19

Species Reactivity: Human. Others not tested.

Epitope: Intracellular region

Immunogen: Synthetic peptide from the intracellular region of CD5

Positive Control: Raji cells, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen CD5
Applications Western Blot
Classification Monoclonal
Clone SP19
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide from the intracellular region of CD5
Quantity 7 mL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Prediluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.