Learn More
Description
Host Species: Mouse
Clone: KP1
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.
Epitope: Not determined
Immunogen: Subcellular fraction of human alveolar macrophages
Molecular Weight: 110 kDa
Positive Control: Tonsil, lymph node, or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD68/Macrophage Marker Ab-3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | KP1 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Subcellular fraction of human alveolar macrophages |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.