Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68/Macrophage Marker Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS397R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Supernatant
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS397R7 7 mL Prediluted -
MS397P 1 mL Concentrated 200μg/mL
MS397RQ 12 mL Supernatant -
MS397PABX 200 μL Concentrated 1mg/mL
MS397P1 500 μL Concentrated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS397R7 Supplier Epredia Supplier No. MS397R7
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

CD68 is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: KP1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.

Epitope: Not determined

Immunogen: Subcellular fraction of human alveolar macrophages

Molecular Weight: 110 kDa

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD68/Macrophage Marker Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KP1
Conjugate Unconjugated
Host Species Mouse
Immunogen Subcellular fraction of human alveolar macrophages
Quantity 7 mL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Feline, Human, Non-human Primate, Rat
Form Prediluted
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.