Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79a/mb-1 (B-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS357P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS357P0 100 μL Unconjugated Concentrated 200μg/mL
MS357P1 500 μL Unconjugated Concentrated 200μg/mL
MS357B0 100 μL Biotin Concentrated 200μg/mL
MS357B1 500 μL Biotin Concentrated 200μg/mL
MS357B 1 mL Biotin Concentrated 200μg/mL
MS357P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS357PABX 200 μL Unconjugated Concentrated 1mg/mL
MS357P 1 mL Unconjugated Concentrated 200μg/mL
MS357R1 1 mL Unconjugated Prediluted Prediluted
MS357R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS357P0 Supplier Epredia Supplier No. MS357P0

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.

Host Species: Mouse

Clone: HM47/A9

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Horse, Cow, Pig, Rabbit, Guinea Pig, Rat, Mouse and Chicken (Plasma cells). Does not react with opossum.

Epitope: GTYQDVGSLNIADVQ

Immunogen: A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein

Molecular Weight: 47 kDa

Positive Control: Raji cells, Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD79a/mb-1 (B-Cell Marker) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HM47/A9
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Bovine, Chicken, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.