Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1171P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1171P1ABX 100 μL Concentrated 1mg/mL
MS1171P0 100 μL Concentrated 200μg/mL
MS1171PABX 200 μL Concentrated 1mg/mL
MS1171P1 500 μL Concentrated 200μg/mL
MS1171R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1171P1ABX Supplier Epredia™ Supplier No. MS1171P1ABX

Ensure accurate, reproducible resuts in immunohistochemistry applications with Epredia™ CD79a/mb-1 (B-Cell Marker) Ab-2, Mouse Monoclonal Antibody.

A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.

Host Species: Mouse

Clone: JCB117

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein

Molecular Weight: 50kDa

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD79a/mb-1 (B-Cell Marker) Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JCB117
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein
Quantity 100 μL
Regulatory Status IVD
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.