Learn More
Description
Host Species: Mouse
Clone: C8/144B
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: C-terminal
Immunogen: A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of α-chain of human CD8 molecule
Molecular Weight: 32 kDa
Positive Control: Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
1. Mason DY, et. al. Journal of Clinical Pathology, 1992, 45(12):1084-8.
Specifications
Specifications
| Antigen | CD8 Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | C8/144B |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of alpha chain of human CD8 molecule |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | T-Cell |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.