Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD8 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS457S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS457S1 500 μL Concentrated
MS457R7 7 mL Prediluted
MS457S0 100 μL Concentrated
MS457S 1 mL Concentrated
MS457R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS457S1 Supplier Epredia™ Supplier No. MS457S1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD8 Ab-1, Mouse Monoclonal Antibody.

CD8 molecule consists of two chains, termed α and Β chain, which are expressed as a disulphide-α β heterdimer or as an alpha α homodimer on T cell subset (cytotoxic/suppressor T cells), thymocytes and NK cells. The majority of CD8+ Tcells expresses CD8 as αβheterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not Β chain).

Host Species: Mouse

Clone: C8/144B

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: C-terminal

Immunogen: A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of α-chain of human CD8 molecule

Molecular Weight: 32 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD8 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C8/144B
Conjugate Unconjugated
Host Species Mouse
Immunogen A 13 amino acid synthetic peptide from the C-terminal cytoplasmic domain of alpha chain of human CD8 molecule
Quantity 500 μL
Regulatory Status IVD
Research Discipline T-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.