Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD8 Rabbit Monoclonal Antibody

Catalog No. RM9116RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9116RQ 12 mL Ready-to-Use
RM9116S1 500 μL Concentrated
RM9116S 1 mL Concentrated
RM9116R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM9116RQ Supplier Epredia™ Supplier No. RM9116RQ

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD8, Rabbit Monoclonal Antibody.

CD8 molecule consists of two chains, termed α and β chain, which are expressed as a disulphide-linked α/β heterdimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 alpha chain (but not β chain).

Host Species: Rabbit

Clone: SP16

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of α-chain of the CD8 molecule

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP16
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of alpha chain of the CD8 molecule
Quantity 12 mL
Regulatory Status IVD
Research Discipline T-Cell
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.