Learn More
CD99/MIC2 (Ewing’s Sarcoma Marker) Ab-2 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: O13
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: Not determined
Immunogen: MeWo human melanoma cells
Molecular Weight: 27-32 kDa
Positive Control: Pancreas or Ewing's sarcoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD99/MIC2 (Ewings Sarcoma Marker) Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | O13 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | MeWo human melanoma cells |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.