Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC25C Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS751P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS751P 1 mL 200μg/mL
MS751P0 100 μL 200μg/mL
MS751P1 500 μL 200μg/mL
MS751P1ABX 100 μL 1mg/mL
MS751PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS751P Supplier Epredia Supplier No. MS751P

CDC25C Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Cyclin-dependent kinases can be activated by CDC25, which removes inhibitory phosphates from tyrosine and threonine residues. At least three CDC25 genes (CDC25A, CDC25B, and CDC25C) have been identified in humans. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.

Host Species: Mouse

Clone: 25C14; same as TC-14

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: aa 1-150

Immunogen: Recombinant human CDC25C protein

Molecular Weight: 53-56 kDa

Positive Control: HeLa, HT29 or LS174T cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CDC25C Ab-1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 25C14; same as TC-14
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human CDC25C protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.