Learn More
Epredia™ Lab Vision™ Cdh1 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: DH01; same as DCS-266
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human Cdh1 protein
Molecular Weight: ∽55 kDa
Positive Control: Jurkat or LS174T cells
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cdh1 Ab-1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | DH01; same as DCS-266 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human cdh1 protein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.