Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cdh1 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1116P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1116P1 500 μL 200μg/mL
MS1116PABX 200 μL 1mg/mL
MS1116P0 100 μL 200μg/mL
MS1116P 1 mL 200μg/mL
MS1116P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1116P1 Supplier Epredia™ Supplier No. MS1116P1

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Epredia™ Cdh1 Ab-1, Mouse Monoclonal Antibody.

Research Areas: Cell Cycle; Cdh1 protein is a limiting, substrate-specific activator of anaphase-promoting complex (APC)-dependent proteolysis and it is required for the degradation of the APC substrates, such as Clb2 and Ase1. Cdh1 plays important role in mitotic regulation.

Host Species: Mouse

Clone: DH01; same as DCS-266

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human Cdh1 protein

Molecular Weight: ∽55 kDa

Positive Control: Jurkat or LS174T cells

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cdh1 Ab-1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DH01; same as DCS-266
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human cdh1 protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.