Learn More
Description
Host Species: Mouse
Clone: DCS-35
Isotype: IgG1/κ
Species Reactivity: Human, Mouse, Rat and Pig. Others not known.
Epitope: aa 1-20
Immunogen: Purified recombinant Cdk4 protein
Molecular Weight: 34 kDa
Positive Control: NIH 3T3, MAD109, LS174T, or MCF-7 cells
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native only)
- Kinase Assay
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cdk4 Ab-1 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | DCS-35 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Purified recombinant cdk4 protein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.