Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk4 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS299P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS299P0 100 μL Unconjugated 200μg/mL
MS299P1 500 μL Unconjugated 200μg/mL
MS299P 1 mL Unconjugated 200μg/mL
MS299PABX 200 μL Unconjugated 1mg/mL
MS299B0 100 μL Biotin 200μg/mL
MS299B1 500 μL Biotin 200μg/mL
MS299B 1 mL Biotin 200μg/mL
MS299P1ABX 100 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS299P0 Supplier Epredia Supplier No. MS299P0

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: DCS-35

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat and Pig. Others not known.

Epitope: aa 1-20

Immunogen: Purified recombinant Cdk4 protein

Molecular Weight: 34 kDa

Positive Control: NIH 3T3, MAD109, LS174T, or MCF-7 cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cdk4 Ab-1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-35
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified recombinant cdk4 protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Mouse, Pig, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.