Learn More
Description
Host Species: Mouse
Clone: DC34
Isotype: IgG1/κ
Species Reactivity: Human, Rat, Mouse and C. elegans. Others not known
Epitope: Not determined
Immunogen: Recombinant Cdk5 protein
Molecular Weight: 31 kDa
Positive Control: HT29, MAD109 or PC12 cells; Brain
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cdk5 Ab-3 |
| Applications | Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | DC34 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant cdk5 protein |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.