Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk5 Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS906P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS906P0 100 μL 200μg/mL
MS906P1 500 μL 200μg/mL
MS906P 1 mL 200μg/mL
MS906P1ABX 100 μL 1mg/mL
MS906PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS906P0 Supplier Epredia Supplier No. MS906P0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Cdk5 Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

Cdk5 is expressed in both cycling and noncycling cells and is present in numerous mammalian tissues. Neuronal and muscle cells contain the highest amount of this protein. The p35 protein, which is expressed solely in the brain, activates Cdk5. Cdk5 activity is involved in terminal differentiation of neurons and muscle cells.

Host Species: Mouse

Clone: DC34

Isotype: IgG1/κ

Species Reactivity: Human, Rat, Mouse and C. elegans. Others not known

Epitope: Not determined

Immunogen: Recombinant Cdk5 protein

Molecular Weight: 31 kDa

Positive Control: HT29, MAD109 or PC12 cells; Brain

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cdk5 Ab-3
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone DC34
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant cdk5 protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species C. elegans, Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.