Learn More
Description
Host Species: Rabbit
Clone: EPR2764Y
Species Reactivity: Human. Others not tested.
Ig Isotype: Rabbit IgG
Epitope: not determined
Immunogen: A synthetic peptide corresponding to residues near the N-terminus of human CDX2
Molecular Weight: 40 kDa
Positive Control: Colon carcinomas
Cellular Localization: Nuclear with occasional cytoplasmic staining on colon carcinoma
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | anti-CDX2 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | EPR2764Y |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to residues near the N-terminus of human CDX2 |
| Quantity | 12 mL |
| Primary or Secondary | Primary |
| Form | Ready-to-Use |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.