Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CDX2 Rabbit Monoclonal Antibody

Catalog No. RM2116S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
7 mL
12 mL
Form:
Ready-to-Use
Supernatant
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2116S 1 mL Supernatant
RM2116R7 7 mL Ready-to-Use
RM2116RQ 12 mL Ready-to-Use
RM2116S0 100 μL Supernatant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM2116S Supplier Epredia™ Supplier No. RM2116S
Add to Cart
Add to Cart
Request a Quote

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CDX2, Rabbit Monoclonal Antibody.

The caudal-related homeodomain protein2 (CDX2) is a transcription factor shown to play a role in the development of small and large intestine in mammals and in the differentiation of intestinal epithelial cells. It has been shown that CDX2 protein detection by IHC coorelates with RNA transcript levels. In studies using 745 cancers from many anatomic sites, colonic adenocarcinomas demonstrated strong staining in 90% of the cases, while adenocarcinomas of the stomach, esophagus and ovary showed extensive staining in only 30% of the cases. Stains of CDX2 and Villin are useful markers in differntial diagnosis of bladder adenocarcinoma and secondary colorectal adenocarcinoma.

Host Species: Rabbit

Clone: EPR2764Y

Species Reactivity: Human. Others not tested.

Ig Isotype: Rabbit IgG

Epitope: not determined

Immunogen: A synthetic peptide corresponding to residues near the N-terminus of human CDX2

Molecular Weight: 40 kDa

Positive Control: Colon carcinomas

Cellular Localization: Nuclear with occasional cytoplasmic staining on colon carcinoma

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen anti-CDX2
Applications Immunohistochemistry
Classification Monoclonal
Clone EPR2764Y
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues near the N-terminus of human CDX2
Quantity 1 mL
Primary or Secondary Primary
Target Species Human
Form Supernatant
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.