Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS613P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS613P0 100 μL Unconjugated Concentrated 200μg/mL
MS613B0 100 μL Biotin Concentrated 200μg/mL
MS613P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS613B1 500 μL Biotin Concentrated 200μg/mL
MS613P1 500 μL Unconjugated Concentrated 200μg/mL
MS613B 1 mL Biotin Concentrated 200μg/mL
MS613R1 1 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS613P0 Supplier Epredia™ Supplier No. MS613P0

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Host Species: Mouse

Clone: COL-1

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Immunogen: Human colon carcinoma extract

Molecular Weight: 180 kDa

Positive Control: LS174T cells, Colon CA

Cellular Localization: Cytoplasmic and luminal membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CEA (Carcinoembryonic Antigen)/CD66e Ab-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone COL-1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human colon carcinoma extract
Quantity 100 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.