Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Chorionic Gonadotropin Human/hCG Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1065P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1065P0 100 μL Concentrated 200μg/mL
MS1065PABX 200 μL Concentrated 1mg/mL
MS1065P1 500 μL Concentrated 200μg/mL
MS1065P 1 mL Concentrated 200μg/mL
MS1065R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1065P0 Supplier Epredia™ Supplier No. MS1065P0

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Mouse

Clone: CG04 + CG05

Isotype: IgG2a/κ + IgG1/κ

Species Reactivity: Human. Others not known

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa (β-hCG)

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CG04 + CG05
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified Beta-subunit of HCG
Quantity 100 μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.