Learn More
Epredia™ Lab Vision™ Chorionic Gonadotropin Human/hCG Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: CG04 + CG05
Isotype: IgG2a/κ + IgG1/κ
Species Reactivity: Human. Others not known
Immunogen: Purified β-subunit of hCG
Molecular Weight: 22 kDa (β-hCG)
Positive Control: Placenta or chorioca
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not optimum)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Chorionic Gonadotropin, Human/hCG Ab-5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CG04 + CG05 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified Beta-subunit of HCG |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.