Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin A Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS381P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS381P 1 mL Concentrated 200μg/mL
MS381P0 100 μL Concentrated 200μg/mL
MS381P1ABX 100 μL Concentrated 1mg/mL
MS381PABX 200 μL Concentrated 1mg/mL
MS381P1 500 μL Concentrated 200μg/mL
MS381R1 1 mL Prediluted -
MS381R7 7 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS381P Supplier Epredia Supplier No. MS381P

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Chromogranin A Ab-2, Mouse Monoclonal Antibody.

Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: PHE5

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with rat. Others not known.

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Chromogranin A Ab-2
Applications Immunofluorescence
Classification Monoclonal
Clone PHE5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human pheochromocytoma
Quantity 1 mL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Non-human Primate
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.