Learn More
Description
Host Species: Mouse
Clone: PHE5
Isotype: IgG1
Species Reactivity: Human and Monkey. Does not react with rat. Others not known.
Immunogen: Human pheochromocytoma
Molecular Weight: 68-75 kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Chromogranin A Ab-2 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | PHE5 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human pheochromocytoma |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Research Discipline | Oncogenesis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.