Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen II Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS234P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS234P0 100 μL 200μg/mL
MS234P1 500 μL 200μg/mL
MS234P1ABX 100 μL 1mg/mL
MS234PABX 200 μL 1mg/mL
MS234P 1 mL 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS234P0 Supplier Epredia Supplier No. MS234P0

Ensure accurate, reproducible results in western blotting and immunofluorescence experiments with Epredia™ Collagen II Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 5B2.5

Isotype: IgG2a/κ

Species Reactivity: Human, Chicken and Mouse. Others not known.

Epitope: GGFDEK, slightly preceding the lysine residue involved in crosslinking. Two terminal glycine residues constitute a key structural element of the epitope. The epitope is lost after digestion of lathyritic type II collagen with recombinant stromelysin.

Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum

Positive Control: Cartilage

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Collagen II Ab-1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5B2.5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Quantity 100 μL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Chicken, Human, Mouse
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.