Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen II Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS235B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS235B1 500 μL Biotin Concentrated 200μg/mL
MS235PABX 200 μL Unconjugated Concentrated 1mg/mL
MS235P1 500 μL Unconjugated Concentrated 200μg/mL
MS235P 1 mL Unconjugated Concentrated 200μg/mL
MS235B0 100 μL Biotin Concentrated 200μg/mL
MS235B 1 mL Biotin Concentrated 200μg/mL
MS235P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS235R1 1 mL Unconjugated Prediluted Prediluted
MS235R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS235B1 Supplier Epredia Supplier No. MS235B1

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.

Host Species: Mouse

Clone: 2B1.5

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.

Epitope: Carboxyl-terminal one quarter of the type II collagen. The epitope for 2B1.5 MAb is not susceptible to digestion of lathyritic type II collagen with recombinant stromelysin

Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum

Positive Control: Cartilage in lung or fetus

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Collagen II Ab-2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B1.5
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Quantity 500 μL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.