Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen VII Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS444P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS444P 1 mL 200μg/mL
MS444P0 100 μL 200μg/mL
MS444P1 500 μL 200μg/mL
MS444P1ABX 100 μL 1mg/mL
MS444PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS444P Supplier Epredia Supplier No. MS444P

Ensure accurate and reproducible results in immunohistochemistry, immunofluorescence, and electron microscopy experiments.

Host Species: Mouse

Clone: LH7.2

Isotype: IgG1/κ

Species Reactivity: Human, Marmoset, Monkey, Cow, Sheep, Goat, Pig and Guinea Pig.

Epitope: On nonhelical (noncollagenous) C-terminal region of collagen VII and is resistant to collagenase, trypsin, pronase, neuraminidase, chondroitinase and hyaluronidase. However, it is lost by digestion of collagen VII with pepsin.

Immunogen: Insoluble fractions of human neonatal foreskin epidermal cells

Molecular Weight: 250 kDa (intact) 150 kDa (digested)

Positive Control: Normal skin

Cellular Localization: Epidermal basement membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen only; fixation in methanol is not suitable)
  • Immunofluorescence
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Collagen VII Ab-1
Applications Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LH7.2
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Insoluble fractions of human neonatal foreskin epidermal cells
Quantity 1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Bovine, Goat, Guinea Pig, Human, Non-human Primate, Pig, Sheep
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.