Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ COX2 Rabbit Monoclonal Antibody

Catalog No. RM9121RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9121RQ 12 mL Ready-to-Use
RM9121S1 500 μL Concentrated
RM9121S 1 mL Concentrated
RM9121R1 1 mL Prediluted
RM9121R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM9121RQ Supplier Epredia™ Supplier No. RM9121RQ

Obtain accurate, reproducible results in Immunohistochemistry experiments with Epredia™ COX2, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio.

COX2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate-early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.

Host Species: Rabbit

Clone: SP21

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from C-terminus of rat COX2

Molecular Weight: 70 kDa

Positive Control: Lung and colon CAs

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen COX2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP21
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide from C-terminus of rat cox2
Quantity 12 mL
Regulatory Status IVD
Research Discipline Enzyme
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.