Learn More
Description
Host Species: Rabbit
Clone: SP21
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide from C-terminus of rat COX2
Molecular Weight: 70 kDa
Positive Control: Lung and colon CAs
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | COX2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP21 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from C-terminus of rat cox2 |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Enzyme |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.