Learn More
Epredia™ Lab Vision™ Cyclin B1 Ab-2 Rabbit Polyclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of human cyclin B1
Molecular Weight: 62 kDa
Positive Control: Jurkat, Ls174T, HeLa cells; Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cyclin B1 Ab-2 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified with BSA and Azide |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide derived from the C-terminus of human cyclin B1 |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.