Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cyclin B1 Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB008P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Formulation:
Purified w/o BSA and Azide
Purified with BSA and Azide
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
RB008P1ABX 500 μL Purified w/o BSA and Azide
RB008P0 100 μL Purified with BSA and Azide
RB008P 1 mL Purified with BSA and Azide
RB008PABX 1 mL Purified w/o BSA and Azide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB008P1ABX Supplier Epredia™ Supplier No. RB008P1ABX

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

In mammals, cyclin B associates with inactive p34cdc2 which facilitates phosphorylation of p34cdc2 at aa 14Thr and 15Tyr. This maintains the inactive state until the end of G2-phase. The inactive cyclin B-p34cdc2 complex continues to accumulate in the cytoplasm until the completion of DNA synthesis, when Cdc25, a specific protein phosphatase, dephosphorylates aa 14Thr and 15Tyr of p34cdc2 rendering the complex active at the G2/M boundary. This mitotic kinase complex remains active until the metaphase/anaphase transition when cyclin B is degraded. This degradation process is ubiquitin-dependent and is necessary for the cell to exit mitosis. So, cyclin B-p34cdc2 plays a critical role in G2 to M transition.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human cyclin B1

Molecular Weight: 62 kDa

Positive Control: Jurkat, Ls174T, HeLa cells; Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cyclin B1 Ab-2
Applications Immunoprecipitation
Classification Polyclonal
Clone Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified w/o BSA and Azide
Host Species Rabbit
Immunogen A synthetic peptide derived from the C-terminus of human cyclin B1
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.