Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cyclin B1 Ab-4 Mouse Monoclonal Antibody, biotin labeled with BSA and azide

Catalog No. MS869R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS869R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS869R1 Supplier Epredia™ Supplier No. MS869R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments.

In mammals, cyclin B associates with inactive p34cdc2. Cyclin B-p34cdc2 plays a critical role in G2 to M transition.

Host Species: Mouse

Clone: GNS11

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat and Hamster. Others not known.

Epitope: aa 1-306

Immunogen: Human cyclin B1 protein

Molecular Weight: 62 kDa

Positive Control: LS174T cells, Tonsil or breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cyclin B1 Ab-4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone GNS11
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Human cyclin B1 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Hamster, Human, Mouse, Rat
Form Prediluted
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.