Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D1 Rabbit Monoclonal Antibody, Epredia™

Catalog No. RM2113R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
7 mL
12 mL
Form:
Ready-to-Use
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2113R1 1 mL Ready-to-Use
RM2113R7 7 mL Ready-to-Use
RM2113RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RM2113R1 Supplier Epredia Supplier No. RM2113R1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cyclin D1, Rabbit Monoclonal Antibody.

Cyclin D1 or PRAD-1 or bcl-1 is a key cell cycle regulator. It functions in association with cdk4 and/or cdk6 by phosphorylating the retinoblastoma protein that releases transcription factors to initiate DNA replication. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. In lymphoid lesions, with the exception of few rare diseases, it is specific for mantle cell lymphoma (MCL).

Host Species: Rabbit

Clone: EPR2241IHC

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1

Molecular Weight: 36 kDa

Positive Control: Mantle cell lymphoma

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Cyclin D1
Applications Immunohistochemistry
Clone EPR2241IHC
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1
Quantity 1 mL
Primary or Secondary Primary
Target Species Human
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.