Learn More
Description
Host Species: Rabbit
Clone: EPR2241IHC
Isotype: IgG
Species Reactivity: Human. Others not tested.
Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1
Molecular Weight: 36 kDa
Positive Control: Mantle cell lymphoma
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Cyclin D1 |
| Applications | Immunohistochemistry |
| Clone | EPR2241IHC |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1 |
| Quantity | 7 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Ready-to-Use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.