Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D2 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS213P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS213P 1 mL 200μg/mL
MS213P0 100 μL 200μg/mL
MS213P1 500 μL 200μg/mL
MS213P1ABX 100 μL 1mg/mL
MS213PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS213P Supplier Epredia Supplier No. MS213P

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation, biological blockade and flow cytometry experiments.

Cyclin D2 is a G1 cyclin required for G1-phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk4 and/or cdk6.

Host Species: Mouse

Clone: DCS-3.1

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D2 protein

Molecular Weight: 34 kDa

Positive Control: U-2-OS, C82, RD, Bristol-8 or LoVo36 cells. Note that T47D, ZR75, SKBR3, MCF-7, BT-20, MDA468, HBL100, BT5549, HeLa, Saos2 and HT29 cells are negative for cyclin D2.

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Biological Blockade
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cyclin D2 Ab-2
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-3.1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human recombinant full length cyclin D2 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.