Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclin D3 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS215R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS215R7 7 mL Prediluted
MS215P0 100 μL Concentrated 200μg/mL
MS215P1 500 μL Concentrated 200μg/mL
MS215P1ABX 100 μL Concentrated 1mg/mL
MS215PABX 200 μL Concentrated 1mg/mL
MS215P 1 mL Concentrated 200μg/mL
MS215R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS215R7 Supplier Epredia Supplier No. MS215R7

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry applications.

Cyclin D3 is a G1 cyclin closely related to cyclin D1 and D2. Cyclins D1, D2, and D3 form active kinase complexes in vivo with a distinct subset of cdks, particularly with cdk4 and cdk6.

Host Species: Mouse

Clone: DCS-22

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human recombinant full-length cyclin D3 protein

Molecular Weight: 34 kDa

Positive Control: LS174T cells, Breast CA or tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cyclin D3 Ab-1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-22
Concentration
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human recombinant full length cyclin D3 protein
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human, Mouse, Non-human Primate, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.