Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1249R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1249R1 1 mL Prediluted Prediluted
MS1249P1 500 μL Concentrated 200μg/mL
MS1249P0 100 μL Concentrated 200μg/mL
MS1249P1ABX 100 μL Concentrated 1mg/mL
MS1249PABX 200 μL Concentrated 1mg/mL
MS1249P 1 mL Concentrated 200μg/mL
MS1249R7 7 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1249R1 Supplier Epredia™ Supplier No. MS1249R1

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and immunoprecipitation experiments.

CFTR (cystic fibrosis transmembrane conductance regulator) is composed of two membrane-spanning domains (MSD), two nucleotide-binding domains (NBD), and an R domain. It is structurally similar to multidrug resistance (Mdr1) protein and both are members of the superfamily of ATP-binding cassette (ABC) transporters, also known as traffic ATPases, which are implicated in the movement of various substrates. The CFTR protein is a small conductance adenosine 3',5'-cyclic monophosphate (cAMP)-activated chloride ion channel found in the apical membranes of epithelia within the pancreas, airway, intestine, bile duct, sweat gland, and male genital ducts. CFTR is a valuable marker of human pancreatic duct cell development and differentiation.

Host Species: Mouse

Clone: M3A7

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1370-1380

Immunogen: Recombinant protein encoding NBF2 domain of human CFTR

Molecular Weight: 165-170 kDa

Positive Control: Pancreas

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Cystic Fibrosis Transmembrane Regulator (CFTR) Ab-4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone M3A7
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Recombinant protein encoding NBF2 domain of human CFTR
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.