Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 10 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS611P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS611P 1 mL Unconjugated Concentrated 200μg/mL
MS611B 1 mL Biotin Concentrated 200μg/mL
MS611B0 100 μL Biotin Concentrated 200μg/mL
MS611B1 500 μL Biotin Concentrated 200μg/mL
MS611P0 100 μL Unconjugated Concentrated 200μg/mL
MS611P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS611PABX 200 μL Unconjugated Concentrated 1mg/mL
MS611P1 500 μL Unconjugated Concentrated 200μg/mL
MS611R1 1 mL Unconjugated Prediluted Prediluted
MS611R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS611P Supplier Epredia Supplier No. MS611P

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

CK10 (Cytokeratin 10) is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.

Host Species: Mouse

Clone: DE-K10

Isotype: IgG1

Species Reactivity: Human, Rat, Dog and Cat. Others not known.

Epitope: Not determined

Immunogen: Cytoskeletal preparation extracted from human ectocervical epithelium

Molecular Weight: 56.5 kDa

Positive Control: A431 cells, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Specifications

Antigen Cytokeratin 10 Ab-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone DE-K10
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Cytoskeletal preparation extracted from human ectocervical epithelium
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Canine, Feline, Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.