Learn More
Description
Host Species: Mouse
Clone: DE-K10
Isotype: IgG1
Species Reactivity: Human, Rat, Dog and Cat. Others not known.
Epitope: Not determined
Immunogen: Cytoskeletal preparation extracted from human ectocervical epithelium
Molecular Weight: 56.5 kDa
Positive Control: A431 cells, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Specifications
| Antigen | Cytokeratin 10 Ab-2 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | DE-K10 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Cytoskeletal preparation extracted from human ectocervical epithelium |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.